MilliporeSigma
  • Conformational peculiarities of alcohols incorporated in lyotropic and thermotropic liquid crystals.
Conformational peculiarities of alcohols incorporated in lyotropic and thermotropic liquid crystals.
Langmuir : the ACS journal of surfaces and colloids (2005-03-19)
Yuji Sasanuma, Futoshi Nishimura, Hiroaki Wakabayashi, Akihiro Suzuki
ABSTRACT
Conformational characteristics of 1-butanol incorporated not only in hexagonal and lamellar aggregates formed by a lyotropic liquid crystal composed of sodium octanoate, 1-butanol, and water but also in a thermotropic liquid crystal, 4'-methoxybenzylidene-4-n-butylaniline (MBBA), have been investigated from 2H NMR quadrupolar splittings of the perdeuterated and partially deuterated compounds. In the lyotropic phases, 1-butanol shows strong trans preferences and renders itself extended, and octanoate decreases the trans fraction toward the methyl terminal to fill the inner space of the aggregate on behalf of 1-butanol. In MBBA, 1-butanol prefers globular conformations. This tendency was also indicated from the phase behaviors of 1-butanol/MBBA and 1-decanol/MBBA systems. In the nematic field, two successive C-C bonds of 1-butanol mostly adopt g+/-g+/- conformations, which are formed in the vicinity of the headgroup of 1-decanol in the lamella of the sodium octanoate/1-decanol/water system. A large degree ofconformational freedom near the hydroxyl group of alcohols, being shown by ab initio molecular orbital calculations at the Gaussian-2 level, permits them to change conformation for each environment.
Sigma-Aldrich
N-(4-Methoxybenzylidene)-4-butylaniline, 98%
Sigma-Aldrich
1-Decanol, 98%
Supelco
1-Decanol, analytical standard
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Decanol, ≥98%