MilliporeSigma
  • Exogenous supply of pantoyl lactone to excised leaves increases their pantothenate levels.
Exogenous supply of pantoyl lactone to excised leaves increases their pantothenate levels.
Annals of botany (2005-03-16)
Bala Rathinasabapathi, Suresh Babu Raman
ABSTRACT
All plants synthesize pantothenate but its synthesis and regulation are not well understood. The aim of this work is to study the effect of exogenous supply of precursor compounds on pantothenate levels in leaves. Precursor compounds were supplied in solution to excised leaves and the pantothenate content was measured using a microbial method. Pantothenate levels in excised leaves of Limonium latifolium, tomato (Lycopersicon esculentum), bean (Phaseolus vulgaris) and grapefruit (Citrus x paradisi) were examined following an exogenous supply of the precursor compounds pantoyl lactone or beta-alanine. Significantly higher levels of extractable pantothenate were found when pantoyl lactone was supplied, but not when beta-alanine was supplied despite a measurable uptake of beta-alanine into the leaf. The results suggested that the pantoate supply may be rate-limiting or regulating pantothenate synthesis in leaves.
Sigma-Aldrich
D-(−)-Pantolactone, 99%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%