MilliporeSigma
  • Degradation of neohesperidin dihydrochalcone by human intestinal bacteria.
Degradation of neohesperidin dihydrochalcone by human intestinal bacteria.
Journal of agricultural and food chemistry (2005-03-03)
Annett Braune, Wolfram Engst, Michael Blaut
ABSTRACT
The degradation of neohesperidin dihydrochalcone by human intestinal microbiota was studied in vitro. Human fecal slurries converted neohesperidin dihydrochalcone anoxically to 3-(3-hydroxy-4-methoxyphenyl)propionic acid or 3-(3,4-dihydroxyphenyl)propionic acid. Two transient intermediates were identified as hesperetin dihydrochalcone 4'-beta-d-glucoside and hesperetin dihydrochalcone. These metabolites suggest that neohesperidin dihydrochalcone is first deglycosylated to hesperetin dihydrochalcone 4'-beta-d-glucoside and subsequently to the aglycon hesperetin dihydrochalcone. The latter is hydrolyzed to the corresponding 3-(3-hydroxy-4-methoxyphenyl)propionic acid and probably phloroglucinol. Eubacterium ramulus and Clostridium orbiscindens were not capable of converting neohesperidin dihydrochalcone. However, hesperetin dihydrochalcone 4'-beta-d-glucoside was converted by E. ramulus to hesperetin dihydrochalcone and further to 3-(3-hydroxy-4-methoxyphenyl)propionic acid, but not by C. orbiscindens. In contrast, hesperetin dihydrochalcone was cleaved to 3-(3-hydroxy-4-methoxyphenyl)propionic acid by both species. The latter reaction was shown to be catalyzed by the phloretin hydrolase from E. ramulus.
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥95% (HPLC)
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥96%, FG
Supelco
Neohesperidin dihydrochalcone, analytical standard