MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Characteristic odor components of kumquat (Fortunella japonica Swingle) peel oil.
Characteristic odor components of kumquat (Fortunella japonica Swingle) peel oil.
Journal of agricultural and food chemistry (2005-03-03)
Hyang-Sook Choi
ABSTRACT
This study was conducted to determine the composition of kumquat (Fortunella japonica Swingle) cold-pressed peel oil and to determine which volatile components are primarily responsible for the aroma of this oil. Eighty-two compounds were identified in the oil by GC and GC-MS. The major compounds were limonene (93.73%), myrcene (1.84%), and ethyl acetate (1.13%). Flavor dilution (FD) factors and relative flavor activities (RFA) of volatile constituents were evaluated by aroma extract dilution analysis with gas chromatography-olfactometry (GC-O). Camphene, terpinen-4-ol, citronellyl formate, and citronellyl acetate showed high FD factors (>/=5) and RFA (>20). Citronellyl formate and citronellyl acetate were regarded as the characteristic odor components of the kumquat peel oil from the results of FD factor, RFA, and GC-sniffing. Citronellyl acetate is considered to be the odor component most similar to kumquat by organoleptic evaluation with GC-O.
Sigma-Aldrich
Citronellyl acetate, ≥95%, FCC, FG