MilliporeSigma
  • Amination of N-aryl prolinol via ring expansion and contraction: application to the chiral ligand for the catalytic asymmetric reaction.
Amination of N-aryl prolinol via ring expansion and contraction: application to the chiral ligand for the catalytic asymmetric reaction.
The Journal of organic chemistry (2005-02-26)
Takashi Mino, Akio Saito, Youichi Tanaka, Shintaro Hasegawa, Yutaka Sato, Masami Sakamoto, Tsutomu Fujita
ABSTRACT
Chiral diaminophosphines 4 were prepared from (S)-prolinol-derived aminophosphine oxide 5 by bromination with ring expansion followed by amination with ring contraction and reduction, using trichlorosilane. In the presence of 4 as a ligand, palladium-catalyzed asymmetric allylic alkylation of 1,3-diphenyl-2-propenyl acetate (11) with a dialkyl malonate-BSA-LiOAc system was successfully carried out with good enantioselectivities (up to 98% ee).
Sigma-Aldrich
(S)-(+)-2-Pyrrolidinemethanol, 97%