MilliporeSigma
  • Multiple isotope effect study of the acid-catalyzed hydrolysis of methyl formate.
Multiple isotope effect study of the acid-catalyzed hydrolysis of methyl formate.
The Journal of organic chemistry (2005-02-26)
John F Marlier, Thomas G Frey, Jennifer A Mallory, W W Cleland
ABSTRACT
Multiple isotope effects have been measured for the acid-catalyzed hydrolysis of methyl formate in 0.5 M HCl at 20 degrees C. The isotope effects in the present investigation include the carbonyl carbon (13k = 1.028 +/- 0.001), the carbonyl oxygen (18k = 0.9945 +/- 0.0009), the nucleophile oxygen (18k = 0.995 +/- 0.001), and the formyl hydrogen ((D)k = 0.81 +/- 0.02). Determination of the carbonyl carbon, carbonyl oxygen, and formyl hydrogen isotope effects was performed via isotopic analysis of residual substrate. However, determination of the oxygen nucleophile isotope effect required analysis of the oxygen atoms of the product (formic acid), which exchange with the solvent (water) under acid conditions. This necessitated measurement of the rate of exchange of these oxygen atoms under the conditions for hydrolysis (k(ex) = 0.0723 min(-1)) and correction of the raw isotope ratios measured during the nucleophile-O isotope effect experiment. These results, along with the previously reported isotope effect for the leaving oxygen (18k = 1.0009) and the ratio of the rate of hydrolysis to that of exchange of the carbonyl oxygen with water (k(h)/k(ex) = 11.3), give a detailed picture of the transition-state structure for the reaction.
Sigma-Aldrich
Methyl formate, reagent grade, 97%
Sigma-Aldrich
Methyl formate, anhydrous, 99%
Supelco
Methyl formate, analytical standard
Sigma-Aldrich
Methyl formate, spectrophotometric grade, ≥98%