MilliporeSigma
  • Substrate modulation of the properties and reactivity of the oxy-ferrous and hydroperoxo-ferric intermediates of cytochrome P450cam as shown by cryoreduction-EPR/ENDOR spectroscopy.
Substrate modulation of the properties and reactivity of the oxy-ferrous and hydroperoxo-ferric intermediates of cytochrome P450cam as shown by cryoreduction-EPR/ENDOR spectroscopy.
Journal of the American Chemical Society (2005-02-03)
Roman Davydov, Roshan Perera, Shengxi Jin, Tran-Chin Yang, Thomas A Bryson, Masanori Sono, John H Dawson, Brian M Hoffman
ABSTRACT
EPR/ENDOR studies have been carried out on oxyferrous cytochrome P450cam one-electron cryoreduced by gamma-irradiation at 77 K in the absence of substrate and in the presence of a variety of substrates including its native hydroxylation substrate, camphor (a), and the alternate substrates, 5-methylenyl-camphor (b), 5,5-difluorocamphor (c), norcamphor (d), and adamantanone (e); the equivalent experiments have been performed on the T252A mutant complexed with a and b. The present study shows that the properties and reactivity of the oxyheme and of both the primary and the annealed intermediates are modulated by a bound substrate. This includes alterations in the properties of the heme center itself (g tensor; (14)N, (1)H, hyperfine couplings). It also includes dramatic changes in reactivity: the presence of any substrate increases the lifetime of hydroperoxoferri-P450cam (2) no less than ca. 20-fold. Among the substrates, b stands out as having an exceptionally strong influence on the properties and reactivity of the P450cam intermediates, especially in the T252A mutant. The intermediate, 2(T252A)-b, does not lose H(2)O(2), as occurs with 2(T252A)-a, but decays with formation of the epoxide of b. Thus, these observations show that substrate can modulate the properties of both the monoxygenase active-oxygen intermediates and the proton-delivery network that encompasses them.
Sigma-Aldrich
2-Adamantanone, ReagentPlus®, 99%