MilliporeSigma
  • The effect of camphorquinone (CQ) and CQ-related photosensitizers on the generation of reactive oxygen species and the production of oxidative DNA damage.
The effect of camphorquinone (CQ) and CQ-related photosensitizers on the generation of reactive oxygen species and the production of oxidative DNA damage.
Biomaterials (2005-01-25)
Dustin Pagoria, Abert Lee, Werner Geurtsen
ABSTRACT
Recent evidence suggests that following visible-light (VL) irradiation, CQ and the CQ-related photosensitizers benzil (BZ), benzophenone (BP), and 9-fluorenone (9-F) generate initiating radicals that may indiscriminately react with molecular oxygen forming reactive oxygen species (ROS). The purpose of this investigation was to determine whether VL-irradiated CQ, BZ, BP, and 9-F cause DNA damage due to the generation of ROS in vitro. ROS formation by CQ and CQ-related photosensitizers+/-dimethyl-p-toluidine (DMT) was investigated in a cell-free system with VL irradiation. DNA damage was determined using PhiX-174 RF I supercoiled double-stranded plasmid DNA and ROS quantified with 4-((9-acridinecarbonyl)amino)-2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO-9-AC), a fluorogenic ROS-sensitive probe. VL-irradiated CQ, BZ, BP, and 9-F (+/-DMT) produced significant DNA damage at 0.1, 0.5, and 1.0 mM and in a concentration-dependent manner (p<0.05). TEMPO-9-AC revealed that all investigated VL-irradiated photosensitizers produced significant amounts of ROS with BZ in the presence of DMT generating the most ROS after 30, 60, and 90 min. VL-irradiated CQ, BZ, BP, and 9-F +/-DMT continued to generate significant amounts of ROS 90 min after VL irradiation. As a result, future investigations should evaluate the effect of VL-irradiated photosensitizers in cells and possible protective effects provided by antioxidants.
Sigma-Aldrich
9-Fluorenone, 98%