MilliporeSigma
  • Protein-assisted synthesis of single-crystal nanowires of bismuth compounds.
Protein-assisted synthesis of single-crystal nanowires of bismuth compounds.
Chemical communications (Cambridge, England) (2005-01-18)
Feng Gao, Qingyi Lu, Sridhar Komarneni
ABSTRACT
Lysozyme, a protein, has been found to be a new morphology-directing agent and a simple and mild bio-molecule assisted method has been proposed for the synthesis of single-crystal bismuth sulfide and oxide nanowires.
Sigma-Aldrich
Bismuth(III) sulfide, 99%