MilliporeSigma
Biocompatible lecithin organogels: structure and phase equilibria.
Langmuir : the ACS journal of surfaces and colloids (2004-12-29)
Ruggero Angelico, Andrea Ceglie, Giuseppe Colafemmina, Francesco Lopez, Sergio Murgia, Ulf Olsson, Gerardo Palazzo
ABSTRACT
The microstructure of organogels formed upon the addition of tiny amounts of water to a solution of lecithin in fatty acid esters (viz. isopropylpalmitate and ethyloleate) was investigated by means of molecular self-diffusion measurements. In both systems lecithin and water form disconnected cylindrical reverse micelles. The ternary phase map for the lecithin/water/isopropylpalmitate has been investigated in detail. The organogel exists in a narrow region close to the lecithin-oil binary axis; for higher water content equilibrium between lamellae and reverse micelles is found. Lamellar phase occupies the lecithin-rich region, close to the lecithin corner (with the exception of a small island of hexagonal phase) and coexists with neat water close to the water-lecithin axis. The remaining part of the phase map shows the three-phase coexistence of water, oil, and lamellar phase.
Sigma-Aldrich
Isopropyl palmitate, ≥90%
Sigma-Aldrich
Isopropyl palmitate, technical grade, 90%
Supelco
Isopropyl palmitate, Pharmaceutical Secondary Standard; Certified Reference Material