MilliporeSigma
  • Synthesis and dynamic NMR study of ketenimines derived from tert-butyl isocyanide, alkyl 2-arylamino-2-oxo-acetates, and dialkyl acetylenedicarboxylates.
Synthesis and dynamic NMR study of ketenimines derived from tert-butyl isocyanide, alkyl 2-arylamino-2-oxo-acetates, and dialkyl acetylenedicarboxylates.
Molecular diversity (2004-12-23)
Issa Yavari, Farough Nasiri, Horieh Djahaniani
ABSTRACT
The adduct produced in the reaction between tert-butyl isocyanide and dialkyl acetylenedicarboxylates was trapped by alkyl 2-arylamino-2-oxo-acetates. When the aryl group is 2-methyl-6-nitrophenyl or 2,6-di-isopropylphenyl, the product exists as two stable rotamers at room temperature as a result of restricted rotation around the Ar-N single bond. When the aryl group is 1-naphthyl or 8-quinolinyl, dynamic NMR effects are observed in the 1H NMR spectra. The calculated free-energy of activation for interconversion of the rotational isomers in 1-naphthyl and 8-quinolinyl derivatives amounts to about 99+/-2 and 68.5+/-2 kJ mol(-1), respectively.
Sigma-Aldrich
tert-Butyl isocyanide, 98%