MilliporeSigma
  • Selective extraction of acetic acid from the fermentation broth produced by Mannheimia succiniciproducens.
Selective extraction of acetic acid from the fermentation broth produced by Mannheimia succiniciproducens.
Biotechnology letters (2004-12-18)
Yun Suk Huh, Yeon Ki Hong, Won Hi Hong, Ho Nam Chang
ABSTRACT
Acetic acid is by-product from fermentation processes for producing succinic acid using Mannheimia succiniciproducens . To obtain pure succinic acid from the final fermentation broth, acetic acid was selectively removed based on the different extractability of succinic acid and acetic acid with pH using tri-n-octylamine (TOA) as extractant. When successive batch extractions were performed using 0.25 mol TOA kg(-1) dissolved in 1-octanol at pH 5, the mol ratio of succinic acid to acetic acid before extraction was 4.9 and the final ratio after the fourth batch was 9.4.
Sigma-Aldrich
Trioctylamine, 98%