MilliporeSigma
  • The long-term effect of toosendanin on current through nifedipine-sensitive Ca2+ channels in NG108-15 cells.
The long-term effect of toosendanin on current through nifedipine-sensitive Ca2+ channels in NG108-15 cells.
Toxicon : official journal of the International Society on Toxinology (2004-12-08)
Mu-Feng Li, Yu-Liang Shi
ABSTRACT
Toosendanin is a triterpenoid derivative extracted from Melia toosendan Sieb et Zucc. Previous studies demonstrated that toosendanin could block neurotransmission and stimulate PC12 cell into differentiation and apoptosis. These actions of toosendanin were suggested to result from a continuous increase in Ca2+ influx, which led to intracellular Ca2+ overload. Here, we observed the long-term effect of toosendanin on Ca2+ channels in NG108-15 cells by whole-cell patch-clamp recording. Obtained data showed that a prolonged exposure to toosendanin induced a continuous increase in the Ca2+ influx in a concentration and time-dependent manner while a brief treatment induced an irreversible increase in Ca2+ influx in differentiated NG108-15 cells. The nifedipine-sensitive L-type currents were significantly increased after exposure to TSN while the nifedipine-resistant or omega-conotoxin MVIIC-sensitive currents were not affected.