MilliporeSigma
  • Structure of the protease domain of memapsin 2 (beta-secretase) complexed with inhibitor.
Structure of the protease domain of memapsin 2 (beta-secretase) complexed with inhibitor.
Science (New York, N.Y.) (2000-10-06)
L Hong, G Koelsch, X Lin, S Wu, S Terzyan, A K Ghosh, X C Zhang, J Tang
ABSTRACT
Memapsin 2 (beta-secretase) is a membrane-associated aspartic protease involved in the production of beta-amyloid peptide in Alzheimer's disease and is a major target for drug design. We determined the crystal structure of the protease domain of human memapsin 2 complexed to an eight-residue inhibitor at 1.9 angstrom resolution. The active site of memapsin 2 is more open and less hydrophobic than that of other human aspartic proteases. The subsite locations from S4 to S2' are well defined. A kink of the inhibitor chain at P2' and the change of chain direction of P3' and P4' may be mimicked to provide inhibitor selectivity.