MilliporeSigma
  • Transformation of 2-aminoacetophenone to ( S) -2-amino-1-phenylethanol by Arthrobacter sulfureus.
Transformation of 2-aminoacetophenone to ( S) -2-amino-1-phenylethanol by Arthrobacter sulfureus.
Biotechnology letters (2004-10-16)
Guogang Zhao, Jianjun Wang, Kai Ma, Liu Yang, Sheng Wu, Yanbin Liu, Wanru Sun
ABSTRACT
A new isolate of Arthrobacter sulfureus , when incubated at 50 g resting cells (dry cell wt) l(-1) with 50 g glucose l(-1) and 1 g 2-aminoacetophenone l(-1) in 50 mm potassium buffer (pH 7, 4 ml) at 30 degrees C, produced ( S )-2-amino-1-phenylethanol (e.e. >99%) with 75% yield in 6 h.
Sigma-Aldrich
2′-Aminoacetophenone, ≥98%
Sigma-Aldrich
2′-Aminoacetophenone, 98%