MilliporeSigma
  • Restoration of ureagenesis in N-acetylglutamate synthase deficiency by N-carbamylglutamate.
Restoration of ureagenesis in N-acetylglutamate synthase deficiency by N-carbamylglutamate.
The Journal of pediatrics (2004-10-14)
Ljubica Caldovic, Hiroki Morizono, Yevgeny Daikhin, Itzhak Nissim, Robert J McCarter, Marc Yudkoff, Mendel Tuchman
ABSTRACT
In a patient with N-acetylglutamate synthase (NAGS) deficiency, incorporation of an isotopic label from ammonium chloride into urea was markedly reduced before treatment with N-carbamyl-L-glutamate (NCLG) and completely normalized following treatment. Blood ammonia rose following ammonium tracer ingestion before treatment but remained low following treatment. Serum urea concentration doubled following the treatment.
Sigma-Aldrich
N-Carbamyl-L-glutamic acid