MilliporeSigma
  • Bacteriorhodopsin-based Langmuir-Schaefer films for solar energy capture.
Bacteriorhodopsin-based Langmuir-Schaefer films for solar energy capture.
IEEE transactions on nanobioscience (2004-09-24)
Paolo Bertoncello, Davide Nicolini, Cristina Paternolli, Valter Bavastrello, Claudio Nicolini
ABSTRACT
The photovoltaic (PV) solar cell, converting incident solar radiation directly into electrical energy, today represents the most common power source for the earth-orbiting spacecraft, and the utilization of organic materials in this context is here explored in comparison with the present state of the art placing emphasis in organic nanotechnology. Poly[3-3'(vinylcarbazole)] (PVK) was synthesized by oxidative polymerization with ferric chloride of N-vinylcarbazole. The resulting polymer was then deposited on solid support by using the Langmuir-Schaefer (LS) technique. The pressure-area isotherm of PVK revealed the possibility of compact monolayer formation at the air-water interface. Different layers of PVK were doped with iodine vapors. The cyclic voltammetry investigation of PVK-doped I2 showed a distinctive electrochemical behavior. The photoinduced charge transfer across a donor/acceptor (D/A) hybrid interface provided an effective method to study the PV properties of the composite LS films. The results are compared with other approaches within the biological framework, such as bacteriorhodopsin (BR), and organic nanostructured materials.
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mw ~1,100,000, powder
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mn 25,000-50,000