MilliporeSigma
  • Dehalogenation of trichlorofluoromethane (CFC-11) by Methanosarcina barkeri.
Dehalogenation of trichlorofluoromethane (CFC-11) by Methanosarcina barkeri.
FEMS microbiology letters (1992-01-01)
U E Krone, R K Thauer
ABSTRACT
Methanobacterium barkeri was found to catalyze the reductive dehalogenation of trichlorofluoromethane (CFC-11), also known as FREON 11. Products detected were CHFCl2, CH2FCl, CO and fluoride.
Sigma-Aldrich
Trichlorofluoromethane solution, NMR reference standard, 10% in acetone-d6 (99.9 atom % D), TMS 6 %, 1,2-dichloro-1,1-difluoroethane 1 %
Sigma-Aldrich
Trichlorofluoromethane, ≥99.5%
Supelco
Trichlorofluoromethane, analytical standard