MilliporeSigma
  • Inhibitory effect of a bitter melon extract on the P-glycoprotein activity in intestinal Caco-2 cells.
Inhibitory effect of a bitter melon extract on the P-glycoprotein activity in intestinal Caco-2 cells.
British journal of pharmacology (2004-09-08)
Tomoko Konishi, Hideo Satsu, Yasuo Hatsugai, Koichi Aizawa, Takahiro Inakuma, Shinji Nagata, Sho-Hei Sakuda, Hiromichi Nagasawa, Makoto Shimizu
ABSTRACT
Extracts of bitter melon, soybean, dokudami and welsh onion by 40% methanol increased the accumulation of rhodamine-123 by Caco-2 cells, suggesting that these extracts inhibited P-glycoprotein (P-gp). The extract of bitter melon was separated in a tC18 cartridge column and the eluate from 80% acetonitrile most markedly increased the [(3)H]-daunomycin accumulation by Caco-2 cells. The inhibitory compounds in the bitter melon fraction were isolated by HPLC with Pegasil C4 and Pegasil ODS columns. The HPLC fraction having the highest activity was analyzed by (1)H-NMR and FAB-MS, and the active compound was identified as 1-monopalmitin. The inhibitory activities of 1-monopalmitin and its related compounds suggested that the inhibition of P-gp activity was not dependent on the degree of unsaturation of fatty acid in the monoglyceride, but on the chain length. It was also suggested that the monoglyceride structure played an important role in the inhibition of P-gp activity. Monoglycerides could therefore alter the pharmacokinetics of drugs by inhibiting the P-gp-mediated efflux.
Sigma-Aldrich
DL-α-Palmitin, ≥99%