MilliporeSigma
  • Potential for use of synthetic sex pheromone for mating disruption of the olive pyralid moth, Euzophera pinguis.
Potential for use of synthetic sex pheromone for mating disruption of the olive pyralid moth, Euzophera pinguis.
Journal of chemical ecology (2004-07-28)
Antonio Ortiz, Antonio Quesada, Adolfo Sanchez
ABSTRACT
The potential for pheromone-based mating disruption of the olive pyralid moth (OPM), Euzophera pinguis, in olive groves was investigated during the second flight period in small-plot trials in 2002. The female of this species emits a blend of (9Z,12E)-tetradecadien-1-ol and (9Z,12E)-tetradecadienyl acetate, which were synthesized for field tests. Mating disruption efficacy in 0.8-ha trials was evaluated using two parameters: reduction of male capture in pheromone traps and reduction of infestation in infestation-prone sites. White rubber septa containing 10 mg of pheromone blend as disruptant were applied at a density of 50 septa/ha for each treatment. Mean catches of E. pinguis males in pheromone traps were greatly reduced (> 95%) in pheromone-treated plots relative to similar traps placed in control plots. In addition, significant reductions were recorded (35-40%) in the oviposition and infestation levels during pheromone treatment. The total amount of pheromone blend released from disruption dispensers during the field trials was estimated to average 5.4 mg/ha/day, over 56 days.
Product Description
Supelco
Myristyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Tetradecanol, Selectophore, ≥99.0%
Sigma-Aldrich
1-Tetradecanol, 97%