MilliporeSigma
  • Adsorption and desorption of lysozyme on nano-sized magnetic particles and its conformational changes.
Adsorption and desorption of lysozyme on nano-sized magnetic particles and its conformational changes.
Colloids and surfaces. B, Biointerfaces (2004-07-21)
Z G Peng, K Hidajat, M S Uddin
ABSTRACT
Adsorption and desorption of lysozyme on nano-sized magnetic particles and its conformational change were studied in this work. Adsorption of lysozyme on nano-sized magnetic particles (Fe(3)O(4)) was carried out at different pH. Maximum adsorption of lysozyme (4.65 mg/m2) occurred at its isoelectric point (pI = 11.1). Differential scanning calorimetry (DSC) results show that the lysozyme adsorbed on magnetic particles did not show any thermal transition over the range 20-100 degrees C. High desorption of lysozyme from magnetic particles was achieved using NaH(2)PO(4) (pH 4.0) (90%) and NaSCN (pH 6.0) (97%) as desorbents. The conformational change of the lysozyme desorbed by NaH(2)PO(4) was small, while the lysozyme desorbed by NaSCN underwent a significant conformational change as measured by the intrinsic fluorescence. Eighty-eight and 82% activity was retained in the desorbed enzyme for desorption by NaH(2)PO(4) and NaSCN, respectively.
Sigma-Aldrich
Sodium thiocyanate solution, BioUltra, 8 M in H2O
Sigma-Aldrich
Sodium thiocyanate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiocyanate, reagent grade, 98-102% (titration)
Sigma-Aldrich
Sodium thiocyanate, ACS reagent, ≥98.0%