MilliporeSigma
  • Unexpected formation of a copper(II) 12-metallacrown-4 with (S)-glutamic-gamma-hydroxamic acid: a thermodynamic and spectroscopic study in aqueous solution.
Unexpected formation of a copper(II) 12-metallacrown-4 with (S)-glutamic-gamma-hydroxamic acid: a thermodynamic and spectroscopic study in aqueous solution.
Dalton transactions (Cambridge, England : 2003) (2004-07-15)
Matteo Tegoni, Francesco Dallavalle, Barbara Belosi, Maurizio Remelli
ABSTRACT
The equilibria of copper(II) with (S)-glutamic-gamma-hydroxamic acid (H2L) were investigated in aqueous solution by different techniques: glass electrode potentiometry; calorimetry; VIS and CD spectrophotometry; and ES-MS. An unexpected pentacopper(II) 12-metallacrown-4 [Cu5L4H(-4)](2-) was detected, analogous to those well known formed by alpha- and beta-aminohydroxamic acids, but of lower stability. Another five species were found: [CuLH]+; [CuL2H2]; [Cu2L2]; [CuL2H]-; and [CuL2]2-. Their structures are proposed based on both spectroscopic and calorimetric data.
Sigma-Aldrich
L-Glutamic acid γ-monohydroxamate