MilliporeSigma
  • Comparative effects of GLP-1 and GIP on cAMP production, insulin secretion, and in vivo antidiabetic actions following substitution of Ala8/Ala2 with 2-aminobutyric acid.
Comparative effects of GLP-1 and GIP on cAMP production, insulin secretion, and in vivo antidiabetic actions following substitution of Ala8/Ala2 with 2-aminobutyric acid.
Archives of biochemistry and biophysics (2004-07-13)
B D Green, V A Gault, P R Flatt, P Harriott, B Greer, F P M O'Harte
ABSTRACT
The two major incretin hormones, glucagon-like peptide-1 (GLP-1), and glucose-dependent insulinotropic polypeptide (GIP), are currently being considered as prospective drug candidates for treatment of type 2 diabetes. Interest in these gut hormones was initially spurred by their potent insulinotropic activities, but a number of other antihyperglycaemic actions are now established. One of the foremost barriers in progressing GLP-1 and GIP to the clinic concerns their rapid degradation and inactivation by the ubiquitous enzyme, dipeptidyl peptidase IV (DPP IV). Here, we compare the DPP IV resistance and biological properties of Abu8/Abu2 (2-aminobutyric acid) substituted analogues of GLP-1 and GIP engineered to impart DPP IV resistance. Whereas (Abu8)GLP-1 was completely stable to human plasma (half-life >12 h), GLP-1, GIP, and (Abu2)GIP were rapidly degraded (half-lives: 6.2, 6.0, and 7.1 h, respectively). Native GIP, GLP-1, and particularly (Abu8)GLP-1 elicited significant adenylate cyclase and insulinotropic activity, while (Abu2)GIP was less effective. Similarly, in obese diabetic (ob/ob) mice, GIP, GLP-1, and (Abu8)GLP-1 displayed substantial glucose-lowering and insulin-releasing activities, whereas (Abu2)GIP was only weakly active. These studies illustrate divergent effects of penultimate amino acid Ala8/Ala2 substitution with Abu on the biological properties of GLP-1 and GIP, suggesting that (Abu8)GLP-1 represents a potential candidate for future therapeutic development.
Sigma-Aldrich
D-2-Aminobutyric acid, 98%
Sigma-Aldrich
DL-2-Aminobutyric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Sigma-Aldrich
L-2-Aminobutyric acid, BioReagent, suitable for cell culture