MilliporeSigma
  • Enantioselective synthesis of alpha-ionone derivatives using an anti S(N)2' substitution of functionalized zinc organometallics.
Enantioselective synthesis of alpha-ionone derivatives using an anti S(N)2' substitution of functionalized zinc organometallics.
Organic letters (2004-07-02)
Darunee Soorukram, Paul Knochel
ABSTRACT
[reaction: see text] The allylic substitution of sterically hindered (2-iodocycloalkyl)phosphates proceeds with complete anti S(N)2' stereoselectivity with mixed diorganozincs of the type RZnCH(2)SiMe(3) in the presence of CuCN.2LiCl. Only the group R of the copper-zinc reagent is transferred in the allylic substitution. This method was used to prepare odoriferous substances such as (R)-alpha-ionone in 97% ee and (R)-dihydro-alpha-ionone in 98% ee.
Sigma-Aldrich
α-Ionone, technical grade, 90%
Sigma-Aldrich
α-Ionone, ≥90%, stabilized