MilliporeSigma
  • Highly efficient biocatalytic resolution of cis- and trans-3-aminoindan-1-ol: syntheses of enantiopure orthogonally protected cis- and trans-indane-1,3-diamine.
Highly efficient biocatalytic resolution of cis- and trans-3-aminoindan-1-ol: syntheses of enantiopure orthogonally protected cis- and trans-indane-1,3-diamine.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-06-24)
Mónica López-García, Ignacio Alfonso, Vicente Gotor
ABSTRACT
The efficient chemoenzymatic synthesis of enantiopure 1,3-difunctionalized indane derivatives has been achieved. The corresponding cis and trans N-protected amino alcohols were successfully resolved by acetylation using lipase B, which is a biocatalyst isolated from Candida antarctica. All the possible isomers were obtained in very good chemical yields and ee values (>99 %). The utility of these compounds was subsequently shown by the preparation of orthogonally protected cis- and trans-indane-1,3-diamine using a Mitsunobu reaction. Both enantiomers of the trans isomer and a desymmetrized cis diastereomer were prepared in enantiopure form. Complete inversion of configuration during the Mitsunobu reaction was demonstrated by a combination of NMR techniques and molecular modeling. The utility and versatility of the strategy was also demonstrated by the selective deprotection of each nitrogen atom under mild reaction conditions.
Sigma-Aldrich
Indan, 95%
Supelco
Indan, analytical standard