MilliporeSigma
  • Ginkgolides, diterpene trilactones of Ginkgo biloba, as antagonists at recombinant alpha1beta2gamma2L GABAA receptors.
Ginkgolides, diterpene trilactones of Ginkgo biloba, as antagonists at recombinant alpha1beta2gamma2L GABAA receptors.
European journal of pharmacology (2004-06-24)
Shelley H Huang, Rujee K Duke, Mary Chebib, Keiko Sasaki, Keiji Wada, Graham A R Johnston
ABSTRACT
Ginkgolides A, B, and C are diterpene trilactones and active constituents of the 50:1 Ginkgo biloba leaf extract widely used in the symptomatic treatment of mild to moderate dementia. Using the two-electrode voltage clamp methodology, these ginkgolides were found to be moderately potent antagonists at recombinant human alpha(1)beta(2)gamma(2L) GABA(A) receptors expressed in Xenopus oocytes. Ginkgolides A, B, and C inhibited the direct action of gamma-aminobutyric acid (GABA) with K(i) values of 14.5+/-1.0, 12.7+/-1.7, and 16.3+/-2.4 microM respectively. Antagonism by these ginkgolides at alpha(1)beta(2)gamma(2L) GABA(A) receptors appears to be noncompetitive as indicated by the nonparallel right shift and reduced maximal GABA response in their GABA concentration-effect curves.
Sigma-Aldrich
Picrotoxinin