MilliporeSigma
  • ydaG and ydbA of Lactococcus lactis encode a heterodimeric ATP-binding cassette-type multidrug transporter.
ydaG and ydbA of Lactococcus lactis encode a heterodimeric ATP-binding cassette-type multidrug transporter.
The Journal of biological chemistry (2004-06-12)
Jacek Lubelski, Piotr Mazurkiewicz, Ronald van Merkerk, Wil N Konings, Arnold J M Driessen
ABSTRACT
Multidrug resistance (MDR)-type transporters mediate the active extrusion of structurally and functionally dissimilar compounds from the cells, thereby rendering cells resistant to a range of drugs. The ydaG and ydbA genes of Lactococcus lactis encode two ATP-binding cassette half-transporters, which both share homology with MDR proteins such as LmrA from L. lactis or the mammalian P-glycoprotein. The ydaG/ydbA genes were cloned and expressed separately and jointly in L. lactis using the nisin-inducible system. When both proteins are co-expressed, several structurally dissimilar drugs such as ethidium, daunomycin, and BCECF-AM are extruded from the cell. YdaG and YdbA could be co-purified as a stable heterodimer. ATPase activity was found to be associated with the YdaG/YdbA heterodimer only and not with the individual subunits. Both the ydaG and ydbA genes are up-regulated in multidrug-resistant L. lactis strains selected for growth in the presence of a variety of toxic compounds. This is the first demonstration of a functional heterodimeric ATP-binding cassette-type MDR transporter.