MilliporeSigma
  • Bioconversion of 3beta-hydroxy-5-cholenoic acid into chenodeoxycholic acid by rat brain enzyme systems.
Bioconversion of 3beta-hydroxy-5-cholenoic acid into chenodeoxycholic acid by rat brain enzyme systems.
Journal of lipid research (2004-06-04)
Nariyasu Mano, Yoshiaki Sato, Masanori Nagata, Takaaki Goto, Junichi Goto
ABSTRACT
We have previously demonstrated that the rat brain contains three unconjugated bile acids, and chenodeoxycholic acid (CDCA) is the most abundantly present in a tight protein binding form. The ratio of CDCA to the other acids in rat brain tissue was significantly higher than the ratio in the peripheral blood, indicating a contribution from either a specific uptake mechanism or a biosynthetic pathway for CDCA in rat brain. In this study, we have demonstrated the existence of an enzymatic activity that converts 3beta-hydroxy-5-cholenoic acid into CDCA in rat brain tissue. To distinguish marked compounds from endogenous related compounds, 18O-labeled 3beta-hydroxy-5-cholenoic acid, 3beta,7alpha-dihydroxy-5-cholenoic acid, and 7alpha-hydroxy-3-oxo-4-cholenoic acid were synthesized as substrates for in vitro incubation studies. The results clearly suggest that 3beta-hydroxy-5-cholenoic acid was converted to 3beta,7alpha-dihydroxy-5-cholenoic acid by microsomal enzymes. The 7alpha-hydroxy-3-oxo-4-cholenoic acid was produced from 3beta,7alpha-dihydroxy-5-cholenoic acid by the action of microsomal enzymes, and Delta4-3-oxo acid was converted to CDCA by cytosolic enzymes. These findings indicate the presence of an enzymatic activity that converts 3beta-hydroxy-5-cholenoic acid into CDCA in rat brain tissue. Furthermore, this synthetic pathway for CDCA may relate to the function of 24S-hydroxycholesterol, which plays an important role in cholesterol homeostasis in the body.