MilliporeSigma
Hydroquinidine therapy in Brugada syndrome.
Journal of the American College of Cardiology (2004-05-18)
Jean-Sylvain Hermida, Isabelle Denjoy, Jérôme Clerc, Fabrice Extramiana, Geneviève Jarry, Paul Milliez, Pascale Guicheney, Stefania Di Fusco, Jean-Luc Rey, Bruno Cauchemez, Antoine Leenhardt
ABSTRACT
We sought to assess hydroquinidine (HQ) efficacy in selected patients with Brugada syndrome (BrS). Management of asymptomatic patients with BrS and inducible arrhythmias remains a key issue. Effectiveness of class Ia antiarrhythmic drugs, which inhibit the potassium transient outward current of the action potential, has been suggested in BrS. From a cohort of 106 BrS patients, we studied 35 who received HQ (32 men; mean age 48 +/- 11 years). Patients had asymptomatic BrS and inducible arrhythmia (n = 31) or multiple appropriate shocks from an implantable cardioverter-defibrillator (ICD) (n = 4). Asymptomatic patients with inducible arrhythmia underwent electrophysiologic (EP)-guided therapy. When ventricular tachycardia (VT)/ventricular fibrillation (VF) inducibility was not prevented, or in case of HQ intolerance, an ICD was placed. Hydroquinidine prevented VT/VF inducibility in 76% of asymptomatic patients who underwent EP-guided therapy. Syncope occurred in two of the 21 patients who received long-term (17 +/- 13 months) HQ therapy (1 syncope associated with QT interval prolongation and 1 unexplained syncope associated with probable noncompliance). In asymptomatic patients who received an ICD (n = 10), one appropriate shock occurred during a follow-up period of 13 +/- 8 months. In patients with multiple ICD shocks, HQ prevented VT/VF recurrence in all cases during a mean follow-up of 14 +/- 8 months. Hydroquinidine therapy prevented VT/VF inducibility in 76% of asymptomatic patients with BrS and inducible arrhythmia, as well as VT/VF recurrence in all BrS patients with multiple ICD shocks. These preliminary data suggest that preventive treatment by HQ may be an alternative strategy to ICD placement in asymptomatic patients with BrS and inducible arrhythmia.
Sigma-Aldrich
Hydroquinine, 98%
Sigma-Aldrich
Hydroquinidine hydrochloride, 98%