MilliporeSigma
  • Danthron inhibits the neurotoxicity induced by various compounds causing oxidative damages including beta-amyloid (25-35) in primary cortical cultures.
Danthron inhibits the neurotoxicity induced by various compounds causing oxidative damages including beta-amyloid (25-35) in primary cortical cultures.
Biological & pharmaceutical bulletin (2004-05-11)
Yong-Soo Kwon, Jae-Young Koh, Dong-Keun Song, Hyoung-Chun Kim, Myung-Sang Kwon, Yeon-Sik Choi, Myung-Bok Wie
ABSTRACT
Oxidative stress caused by an elevation in reactive oxygen species (ROS) plays an important role in Alzheimer's disease and other neurodegenerative diseases. In this study, we examined the neuroprotective effect of danthron (1,8-dihydroxyanthraquinone) against neurotoxicities induced by beta-amyloid (25-35), excitotoxins, apoptosis, and oxidative stress in primary cortical cultures. Danthron dose-dependently reduced neuronal injury induced by 30 microM beta-amyloid (25-35). Danthron significantly inhibited oxidative injury induced by 100 microM Fe(3+) and decreased membrane lipid peroxidation induced by 100 microM Fe(3+) as measured by thiobarbituric-acid-reactive substance (TBARS). Danthron (0.5-50 microM) ameliorated the effects of buthionine sulfoximine (BSO, 1 mM), which depletes endogenous glutathione by 10-73%. Danthron also dose-dependently inhibited neuronal injury mediated by nitric oxide (NO) radicals, but failed to inhibit injury due to superoxide radicals (O(2-)). These results suggest danthron treatment may, in part, reduce neurotoxicity related to beta-amyloid protein by both dominant inhibitory effects on membrane lipid peroxidation and glutathione deprivation.