MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Simultaneous determination of E-2-nonenal and beta-damascenone in beer by reversed-phase liquid chromatography with UV detection.
Simultaneous determination of E-2-nonenal and beta-damascenone in beer by reversed-phase liquid chromatography with UV detection.
Journal of chromatography. A (2004-04-07)
L F Guido, J R Carneiro, J R Santos, P J Almeida, J A Rodrigues, A A Barros
ABSTRACT
A method for the simultaneous determination of E-2-nonenal and beta-damascenone in beer by reversed-phase liquid chromatography using UV detection is presented. The method consists of beer steam distillation, followed by an extraction/concentration step using Sep-Pak Plus C18 RP cartridges and determination by HPLC at 226 nm UV-absorption maximum. The identity of the compounds was confirmed by GC analysis with MS detection of the isolated fractions. A recovery factor of approximately 80% was obtained for beta-damascenone with a R.S.D. of 3%. E-2-Nonenal and beta-damascenone were monitored in a comparative study of fresh and either naturally and forced aged beer. The results obtained show that both compounds have a similar behaviour through an extended storage of beer and consequently can be used as good analytical markers of beer ageing. Nevertheless, the use of beta-damascenone seems to be more convenient because this compound appears in beer in higher concentrations than E-2-nonenal, thus making it easier to measure.
Sigma-Aldrich
trans-2-Nonenal, 95%
Sigma-Aldrich
Damascenone, natural, 1.1-1.4 wt. % (190 proof ethanol), FG