MilliporeSigma
  • Degradation of ethyl docosahexaenoate by gamma-ray irradiation and suppression of this degradation by antioxidants.
Degradation of ethyl docosahexaenoate by gamma-ray irradiation and suppression of this degradation by antioxidants.
Bioscience, biotechnology, and biochemistry (2004-04-02)
Yoshiyuki Murata, Ken-ichi Osaki, Yasuaki Shimoishi, Naomichi Baba, Mikiro Tada
ABSTRACT
Ethyl 4,7,10,13,16,19-docosahexaenoate (DHA-Et) dissolved in an ethanol solution or embedded in liposomes was degraded by irradiating with gamma rays in a dose-dependent manner. The degradation rate of DHA-Et embedded in liposomes was higher than that of DHA-Et dissolved in ethanol. Antioxidants suppressed the degradation of DHA-Et embedded in liposomes, the order of activity of the antioxidants being luteolin>fisetin>kaempferol>quercetin>rutin. These results suggest that the hydrophobicity (logP) of an antioxidant is one of determinants for antioxidative activity, but that a vicinal diol structure in the B ring is not favorable for the antioxidative activity.
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid ethyl ester, ≥97%
Docosahexaenoic acid ethyl ester, European Pharmacopoeia (EP) Reference Standard