MilliporeSigma
  • A new type of sample tube for reducing convection effects in PGSE-NMR measurements of self-diffusion coefficients of liquid samples.
A new type of sample tube for reducing convection effects in PGSE-NMR measurements of self-diffusion coefficients of liquid samples.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2004-03-26)
Kikuko Hayamizu, William S Price
ABSTRACT
Pulsed gradient spin-echo (PGSE) NMR measurements of the self-diffusion coefficients of low viscosity liquids are greatly hampered by the effects of convection especially away from ambient temperature. Here we report on a new NMR tube designed to minimize the deleterious effects of convection. In this tube, which derives from a Shigemi symmetrical NMR tube, the sample is contained in an annulus formed from a concentric cylinder of susceptibility matched glass. The performance of this tube was demonstrated by conducting measurements on the electrochemically important LiN(SO3CF3)2 (LiTFSI)-diglyme (DG) system. Calibrations were first made using DG at column heights of 2, 3, and 4-mm in the temperature range between -40 and 100 degrees C. Measurements of the diffusion coefficients of the lithium, anion, and DG were then performed to probe the solvent-ion and ion-ion interactions in the DG doped with LiTFSI. Changes in the 1H, 7Li, and 19F PGSE-NMR attenuation curves at -40 degrees C provided clear evidence of interactions between the DG and lithium ion.
Product Description
Supelco
Diethylene glycol dimethyl ether, analytical standard
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%
Sigma-Aldrich
Diethylene glycol dimethyl ether, ReagentPlus®, 99%