MilliporeSigma
  • Adipostatins A and B, new inhibitors of glycerol-3-phosphate dehydrogenase.
Adipostatins A and B, new inhibitors of glycerol-3-phosphate dehydrogenase.
The Journal of antibiotics (1992-06-01)
N Tsuge, M Mizokami, S Imai, A Shimazu, H Seto
ABSTRACT
As a result of screening for inhibitors of glycerol-3-phosphate dehydrogenase, which may be effective to prevent corpulence, we isolated two inhibitors named adipostatin A and adipostatin B from the culture broth of Streptomyces cyaneus 2299-SV1. Their structures have been established to be 5-n-pentadecylresorcinol and 5-isopentadecylresorcinol, respectively. Adipostatin A and adipostatin B inhibited glycerol-3-phosphate dehydrogenase at the IC50 values of 4.1 microM and 4.5 microM, respectively. These compounds prevented triglyceride accumulation in 3T3-L1 cells at a concentration of the microM level.
Product Description
Supelco
5-Pentadecylresorcinol, analytical standard