MilliporeSigma
Effect of ibuprofen on bilirubin-albumin binding.
The Journal of pediatrics (2004-03-06)
Charles E Ahlfors
ABSTRACT
Ibuprofen is used for closing the ductus arteriosus in premature newborn infants. Ibuprofen interferes with bilirubin-albumin binding and increases the unbound bilirubin in pooled newborn plasma to levels similar to those produced by sulfisoxazole, a drug that causes kernicterus in premature newborn infants.
Product Description
Supelco
Sulfisoxazole, VETRANAL®, analytical standard