MilliporeSigma
  • Application of shielded column liquid chromatography for determination of sulfamonomethoxine, sulfadimethoxine, and their N4-acetyl metabolites in milk.
Application of shielded column liquid chromatography for determination of sulfamonomethoxine, sulfadimethoxine, and their N4-acetyl metabolites in milk.
Journal of chromatography. A (2004-02-19)
Kunihiro Kishida, Naoto Furusawa
ABSTRACT
A hazardous-chemical free method for simultaneous determination of sulfamonomethoxine (SMM), sulfadimethoxine (SDM), and their N4-acetyl metabolites in raw milk using shielded column liquid chromatography is developed. The target analytes are extracted by mixing with ethanol-acetic acid (97:3, v/v) followed by centrifugation. The procedure uses a Hisep shielded hydrophobic phase (SHP) column, isocratic elution with 0.1% acetic acid solution (pH 3.1, in water)-ethanol (75:25, v/v), and a photo-diode array detector. Average recoveries from samples spiked at 25-500 ng/ml for each drug were >81% with relative standard deviations within 5%. The limits of quantitation were <25 ng/ml.
Sigma-Aldrich
Sulfamonomethoxine
Supelco
Sulfamonomethoxine, VETRANAL®, analytical standard