MilliporeSigma
  • Preparation of new nitrogen-bridged heterocycles. 55. Reinvestigation of the bromination/dehydrobromination of ethyl 3-[2-(methylthio)indolizin-3-yl]acrylate derivatives.
Preparation of new nitrogen-bridged heterocycles. 55. Reinvestigation of the bromination/dehydrobromination of ethyl 3-[2-(methylthio)indolizin-3-yl]acrylate derivatives.
Chemical & pharmaceutical bulletin (2004-02-06)
Akikazu Kakehi, Suketaka Ito, Hiroyuki Suga, Yuhichi Kobayashi, Peng Zuo, Ryozo Iriye
ABSTRACT
The bromination/dehydrobromination reactions of ethyl 3-[1-alkoxycarbonyl-2-(methylthio)indolizin-3-yl]acrylates were reinvestigated. Reactions of the title compounds with two equivalents of bromine, followed by heating of the resulting reaction mixture and then treatment with a base gave the unexpected dialkyl 7-bromothieno[2,3-b]indolizine-2,9-dicarboxylates, while similar reactions using benzyltrimethylammonium tribromide as a brominating agent afforded only non-brominated thieno[2,3-b]indolizine derivatives, which were converted to the corresponding 7-bromo derivatives upon further treatment with bromine.
Sigma-Aldrich
Benzyltrimethylammonium bromide, 97%
Sigma-Aldrich
Benzyltrimethylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Benzyltrimethylammonium chloride solution, technical, ~60% in H2O
Sigma-Aldrich
Benzyltrimethylammonium hydroxide solution, 40 wt. % in methanol
Sigma-Aldrich
Benzyltrimethylammonium chloride, 97%