MilliporeSigma
  • Diastereoselective additions of chiral vinylzinc reagents to alpha-chiral aldehydes.
Diastereoselective additions of chiral vinylzinc reagents to alpha-chiral aldehydes.
Organic letters (2004-01-30)
James A Marshall, Patrick Eidam
ABSTRACT
[reaction: see text] Additions of vinylic zinc bromide reagents to alpha-chiral aldehydes (R(1) = CH(2)OTBS, R(2) = Me; R(1) = Me, R(2) = OTBS) in the presence of lithiated (+)- or (-)-N-methylephedrine proceed with predominant reagent control to afford anti or syn adducts stereoselectively, except when the aldehydes possess an alkoxy substituent at the alpha- or beta-positions (R(1) = Me, R(2) = OBn; R(1) = CH(2)OBn, R(2) = Me), in which case chelation-controlled adducts predominate.
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF