MilliporeSigma
  • Vibrational spectrum, ab initio calculations, conformational equilibria and torsional modes of 1,3-dichloropropane.
Vibrational spectrum, ab initio calculations, conformational equilibria and torsional modes of 1,3-dichloropropane.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2004-01-30)
Daniel J Duffy, Jason Quenneville, T M Baumbaugh, S A Kitchener, R K McCormick, C N Dormady, T A Croce, A Navabi, Howard D Stidham, Shaw L Hsu, Gamil A Guirgis, Shiping Deng, James R Durig
ABSTRACT
Ab initio calculations are reported for three of four possible conformers of 1,3-dichloropropane. The fourth conformer, with Cs symmetry, has a predicted enthalpy difference of more than 1500 cm(-1) from the most stable conformer from each calculation regardless of the basis set used, so there is little chance of observing it. Thus, there is no evidence in the infrared or Raman spectrum of the presence of a fourth conformer. The order of stability given by the ab initio calculations is C2(GG)>C1(AG)>C2v(AA)>Cs(GG'), where A indicates the anti form for one of the CH2Cl groups and G indicates the gauche conformation for the other CH2Cl group relative to the plane of the carbon atoms. Almost every band observed can be confidently assigned to one or another of the conformers. Many observed bands proved to be of a composite nature, with several nearly coincident vibrations of different conformers contributing to the band contour. Nonetheless, a complete assignment of fundamentals is possible for the most stable C2 conformer, and 5 of the fundamentals of the C2v conformer and 13 those of the C1 conformer can be confidently assigned.
Sigma-Aldrich
1,3-Dichloropropane, 99%