MilliporeSigma
  • Degradation of alpha-pinene oxide and [2H7]-2,5,6-trimethyl-hept-(2E)-enoic acid by Pseudomonas fluorescens NCIMB 11761.
Degradation of alpha-pinene oxide and [2H7]-2,5,6-trimethyl-hept-(2E)-enoic acid by Pseudomonas fluorescens NCIMB 11761.
Journal of biotechnology (2004-01-23)
H Zorn, F Neuser, R G Berger
ABSTRACT
When submerged cultured Pseudomonas fluorescens NCIMB 11761 was fed-batch supplemented with alpha-pinene oxide, a rapid formation of 2,6-dimethyl-5-methylene-hept-(2Z)-enal (I) (isonovalal) was observed. Biotransformation and isomerisation of (I) to the (2E)-isomer (II) (novalal) were enhanced by Lewatit OC 1064, a macroporous polystyrene adsorbent. Accelerated isomerisation in the presence of an amino donor (glycine) at pH 7.3 pointed to a merely chemical mechanism. A maximum yield of 48 g of aldehydesl(-1) was achieved, but quantitative analysis of the volatile fraction showed that the molar conversion of the pinene oxide substrate reached no more than 67%. To fill this gap of the mass balance, the acidic fraction was isolated. It contained several compounds which suggested a beta-oxidation-like catabolism starting from 2,6-dimethyl-5-methylene-hept-(2E)-enoic acid (III) (novalic acid). Using [2H7]-2,5,6-dimethyl-hept-(2E)-enoic acid as a conversion substrate and gas chromatography coupled to atomic emission detection and mass spectrometry a degradation pathway via labelled 3,4-dimethylpentenoic and methylpropanoic acids was evidenced. This pathway may play a predominant role in isoprenoid degradation by soil bacteria.
Sigma-Aldrich
α-Pinene oxide, 97%
Pricing and availability is not currently available.