MilliporeSigma
  • Stereoselective diversity-oriented solution and solid-phase synthesis of tetrahydroquinoline-based polycyclic derivatives.
Stereoselective diversity-oriented solution and solid-phase synthesis of tetrahydroquinoline-based polycyclic derivatives.
Journal of combinatorial chemistry (2004-01-13)
Prabhat Arya, Patricia Durieux, Zai-Xin Chen, Reni Joseph, Donald M Leek
ABSTRACT
A diversity-oriented solution and solid-phase synthesis of tetrahydroquinoline-based tricyclic derivatives has been achieved from enantiomerically pure, natural product-like bicyclic scaffold. The solution synthesis of enantiopure bicyclic scaffold was developed by asymmetric hetero Michael reaction. Our approach for the synthesis of polycyclic derivatives utilized regio- and stereoselective hetero Michael reaction and ring-closing metathesis as key steps in solution and on solid phase.
Sigma-Aldrich
Lithium borohydride, hydrogen-storage grade, ≥90%
Sigma-Aldrich
Lithium borohydride, ≥90%
Sigma-Aldrich
Lithium borohydride, ≥95.0%
Sigma-Aldrich
Lithium borohydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium borohydride solution, 0.5 M in diethyl ether