MilliporeSigma
  • Comparative methyl linoleate and methyl linolenate oxidation in the presence of bovine serum albumin at several lipid/protein ratios.
Comparative methyl linoleate and methyl linolenate oxidation in the presence of bovine serum albumin at several lipid/protein ratios.
Journal of agricultural and food chemistry (2004-01-07)
Rosario Zamora, Francisco J Hidalgo
ABSTRACT
The oxidation of methyl linoleate (LMe) and methyl linolenate (LnMe) in the presence of bovine serum albumin (BSA) in the dark at 60 degrees C was studied to analyze the role of the type of fatty acid and the protein/lipid ratio on the relative progression of the processes involved when lipid oxidation occurs in the presence of proteins. The disappearance of the fatty acid, the formation of primary and secondary products of lipid peroxidation, the loss of amino acid residues, the production of oxidized lipid/amino acid reaction products, and the development of color and fluorescence were studied as a function of incubation time in protein/lipid samples at 10:1, 6:1, and 3:1 w/w ratios. The incubation of LMe and LnMe in the presence of BSA at 60 degrees C rapidly produced lipid peroxidation and protein damage. Although reaction rates were much faster for LnMe than for LMe, both fatty acids had similar behaviors, and LnMe seemed to be only slightly more reactive than LMe for BSA by producing a higher increase of protein pyrroles in the protein and the development of increased browning and fluorescence. The protein/lipid ratio also influenced the relative progress of the reactions implicated. Thus, a lower protein/lipid ratio increased sample oxidation and protein damage. This also produced an increased browning, in accordance with the mechanisms proposed for browning production by oxidized lipid/protein reactions. On the contrary, browning of extracted lipids increased at higher protein/lipid ratios. This opposite tendency allowed evaluation of the overall significance of the different browning processes implicated in the final colors observed, concluding that color changes observed in BSA/lipid samples were mostly a consequence of oxidized lipid/protein reactions.
Sigma-Aldrich
Methyl linolenate, ≥99% (GC)
Supelco
Methyl linolenate, analytical standard
Sigma-Aldrich
Methyl linolenate, technical, 70-80% (GC)