MilliporeSigma
  • Fungal aerobic reductive dechlorination of ethyl 2-chloroacetoacetate by Saccharomyces cerevisiae: mechanism of a novel type of microbial dehalogenation.
Fungal aerobic reductive dechlorination of ethyl 2-chloroacetoacetate by Saccharomyces cerevisiae: mechanism of a novel type of microbial dehalogenation.
Chembiochem : a European journal of chemical biology (2003-12-26)
Gerhard Jörg, Martin Bertau
ABSTRACT
Saccharomyces cerevisiae reduces the beta-keto ester ethyl 2-chloroacetoacetate to the respective chiral cis- and trans-beta-hydroxy esters. In the course of chiral reduction, competing dehalogenation of the xenobiotic substrate to ethyl acetoacetate occurs, in a reaction mediated by cytosolic glutathione (GSH). Mechanistically, the dechlorination is a novel type of glutathione-dependent dehalogenation catalysed by an as yet unidentified glutathione-dependent dehalogenase. The first step consists of a nucleophilic replacement of the chloride substituent by glutathione. In the subsequent enzyme-catalysed step, a second glutathione molecule liberates the dehalogenation product by thiolytic attack at the thioether bridge, resulting in a net transfer of two electrons to the substrate and in the formation of glutathione disulfide (GSSG). Being effective under aerobic conditions and catalysed by a fungus, this reductive dechlorination of an aliphatic substrate is an outstanding example of a novel, glutathione-mediated microbial dehalogenation.
Sigma-Aldrich
Ethyl 4-chloroacetoacetate, 95%
Sigma-Aldrich
Ethyl 4-chloroacetoacetate, Arxada quality, ≥97.0% (GC)