MilliporeSigma
  • Determination of low molecular weight aliphatic primary amines in urine as their benzenesulphonyl derivatives by gas chromatography with flame photometric detection.
Determination of low molecular weight aliphatic primary amines in urine as their benzenesulphonyl derivatives by gas chromatography with flame photometric detection.
Biomedical chromatography : BMC (1992-09-01)
H Kataoka, S Ohrui, Y Miyamoto, M Makita
ABSTRACT
A selective and sensitive method for the determination of low molecular weight aliphatic primary amines in urine is described. These amines were converted into their benzenesulphonyl derivatives by a modified Hinsberg procedure, and measured by gas chromatography with flame photometric detection (FPD-GC) using a DB-1 capillary column. The derivatives were very stable and provided excellent FPD responses. By FPD-GC, linear calibration curves were obtained in the range 10-200 ng of methylamine, ethylamine, n-propylamine, isobutylamine and n-butylamine using tert-butylamine as an internal standard, and the detection limits of these amines were ca. 6-25 pg as the injection amount. Benzenesulphonamide derived from ammonia was converted into its N-dimethylaminomethylene derivative which has a longer retention time, and separated from benzenesulphonyl derivatives of low molecular weight primary amines on the chromatogram. The recoveries of aliphatic primary amines added to urine samples were 91-107% and the relative standard deviations were 0.2-4.5%. Analytical results of aliphatic primary amine contents in urine samples of normal subjects are presented.
Sigma-Aldrich
Benzenesulfonyl chloride, 96%
Sigma-Aldrich
Benzenesulfonyl chloride, for HPLC derivatization
Sigma-Aldrich
Benzenesulfonyl chloride, 99%