MilliporeSigma
  • 3-Aminophenol as a novel coupling agent for the spectrophotometric determination of sulfonamide derivatives.
3-Aminophenol as a novel coupling agent for the spectrophotometric determination of sulfonamide derivatives.
Farmaco (Societa chimica italiana : 1989) (2003-11-25)
P Nagaraja, H S Yathirajan, C R Raju, R A Vasantha, P Nagendra, M S Hemantha Kumar
ABSTRACT
A rapid, simple and sensitive spectrophotometric method for the determination of some sulfa drugs is described. The method is based on the formation of orange yellow colored azo product by the diazotization of sulfonamides, viz., dapsone (DAP), sulfathiazole (SFT), sulfadiazine (SFD), sulfacetamide (SFA), sulfamethoxazole (SFMx), sulfamerazine (SFMr), sulfaguanidine (SFG) and sulfadimidine (SFDd) followed by a coupling reaction with 3-aminophenol in aqueous medium. Absorbance of the resulting orange yellow product is measured at 460 nm and is stable for 6 days at 27 degrees C. Beer's law is obeyed in the concentration range of 0.05-8.0 microg/ml at the wavelength of maximum absorption. The method is successfully employed for the determination of sulfonamides in various pharmaceutical preparations and common excipients used as additives in pharmaceuticals do not interfere in the proposed method. Plausible reaction mechanism is proposed for the formation of the azo product.
Sigma-Aldrich
3-Aminophenol, 98%
Supelco
3-Aminophenol, PESTANAL®, analytical standard