MilliporeSigma
[Preparation of Harris hematoxylin without use of mercury oxide].
Morfologiia (Saint Petersburg, Russia) (2003-11-25)
D E Korzhevskiĭ
ABSTRACT
The study was carried out with the aim of improvement of Harris' alum hematoxylin formula. In presented modified formula toxic mercuric oxide (2.5 g) is substituted by potassium iodate (0.55 g). Alum hematoxylin solution prepared according to suggested formula retains high quality of original Harris' hematoxylin.
Sigma-Aldrich
Mercury(II) oxide yellow, ACS reagent, ≥99.0%
Sigma-Aldrich
Mercury(II) oxide, 99.999% trace metals basis