MilliporeSigma
  • Diversity of L-leucine catabolism in various microorganisms involved in dairy fermentations, and identification of the rate-controlling step in the formation of the potent flavour component 3-methylbutanal.
Diversity of L-leucine catabolism in various microorganisms involved in dairy fermentations, and identification of the rate-controlling step in the formation of the potent flavour component 3-methylbutanal.
Applied microbiology and biotechnology (2003-11-19)
B A Smit, W J M Engels, J T M Wouters, G Smit
ABSTRACT
Various microorganisms, belonging to the genera Lactococcus, Lactobacillus, Streptococcus, Leuconostoc, Bifidobacterium, Propionibacterium, Brevibacterium, Corynebacterium and Arthrobacter, used in dairy fermentations such as cheese making, were analysed for their potential to convert leucine into flavour components, most notably 3-methylbutanal. A large variation between and within species was observed for various enzyme activities involved in the conversion pathway, e.g. transaminases, alpha-hydroxy acid dehydrogenase and alpha-keto acid decarboxylase. In particular, alpha-keto acid decarboxylase activity-leading to 3-methylbutanal-was found to be present in only two of the strains tested. It is proposed that this activity is rate-controlling in the conversion pathway leading to the flavour compound 3-methylbutanal.
Sigma-Aldrich
Isovaleraldehyde, natural, ≥95%, FG
Sigma-Aldrich
Isovaleraldehyde, ≥97%, FG
Sigma-Aldrich
Isovaleraldehyde, 97%