MilliporeSigma
  • Synthetic studies toward FR182877. Remarkable solvent effect in the vinylogous morita-baylis-hillman cyclization.
Synthetic studies toward FR182877. Remarkable solvent effect in the vinylogous morita-baylis-hillman cyclization.
Organic letters (2003-10-24)
Joey L Methot, William R Roush
ABSTRACT
[reaction: see text]. The intramolecular vinylogous Morita-Baylis-Hillman reaction was explored to access the central cyclopentane ring of FR182877. The reaction manifold and product distribution is strikingly solvent and substrate dependent.
Sigma-Aldrich
Trimethylphosphine, 97%
Sigma-Aldrich
Trimethylphosphine solution, 1.0 M in THF
Sigma-Aldrich
Trimethylphosphine solution, 1.0 M in toluene