MilliporeSigma
  • Suppression of intestinal smooth muscle contraction by phenolic compounds.
Suppression of intestinal smooth muscle contraction by phenolic compounds.
Research communications in chemical pathology and pharmacology (1992-09-01)
N Ogata, M Toyoda, T Shibata
ABSTRACT
Wood creosote, a mixture of guaiacol, creosol, and other related phenolic compounds, suppresses the spontaneous longitudinal phasic contractions of an isolated rat ileal segment. Thirty-two phenolic compounds were screened for this activity to identify the active substances in wood creosote. Of the 32 compounds tested, 2,3,6-trimethylphenol, 2,4,6-trimethylphenol, and 4-ethylguaiacol were most effective and their respective IC50 values were 0.035, 0.037, and 0.043 mM. Guaiacol and creosol, the chief constituents of wood creosote, were moderately effective and their respective IC50 values were 0.18 and 0.13 mM. Other phenolic compounds in wood creosote also suppressed the ileal contraction although their IC50 values differed markedly from each other. These results suggest that practically all of its constituent phenolic compounds contribute to wood creosote's capacity to suppress ileal contraction.
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%, FG
Sigma-Aldrich
2-Methoxy-4-methylphenol, natural, 97%, FG
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%