MilliporeSigma
  • Stereospecific hydroxylation of (+)-cedrol by using human skin microbial flora Staphylococcus epidermidis.
Stereospecific hydroxylation of (+)-cedrol by using human skin microbial flora Staphylococcus epidermidis.
Natural product research (2003-10-07)
Mitsuo Miyazawa, Yumi Itsuzaki, Keiji Ishikawa, Kaname Ishisaka
ABSTRACT
Microbial transformation of (+)-cedrol was investigated by using Staphylococcus epidermidis and found that stereospecific hydroxylation of (+)-cedrol occurred at the C-3 position to form (+)-(3S)-3-hydroxycedrol.
Sigma-Aldrich
(+)-Cedrol, ≥99.0% (sum of enantiomers, GC)